Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006928.2 | NP_001006929.1 | 3 - 4 | 522 | 100 | |
GenBank mRNA | BC077032.1 | - | 3 - 4 | 526 | 100 | |
CR760177.2 | - | 3 - 4 | 492 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |