Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007867.1 | NP_001007868.1 | 4 - 5 | 356 | 97 | |
XM_012956794.2 | XP_012812248.1 | 4 - 5 | 442 | 97 | ||
GenBank mRNA | BC080132.1 | - | 4 - 5 | 356 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |