Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007898.1 | NP_001007899.1 | 3 - 4 | 654 | 75 | |
GenBank mRNA | BC080334.1 | - | 3 - 4 | 654 | 75 | |
CR760379.1 | - | 3 - 4 | 616 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |