Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007940.1 | NP_001007941.1 | 3 - 4 | 443 | 74 | |
XM_018093014.1 | XP_017948503.1 | 3 - 4 | 311 | 74 | ||
XM_018093015.1 | XP_017948504.1 | 3 - 4 | 512 | 74 | ||
GenBank mRNA | BC080448.1 | - | 3 - 4 | 443 | 74 | |
CR761604.2 | - | 3 - 4 | 454 | 74 | ||
CT025478.2 | - | 3 - 4 | 452 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |