Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007949.2 | NP_001007950.1 | 1 - 2 | 583 | 90 | |
GenBank mRNA | BC080457.1 | - | 1 - 2 | 587 | 90 | |
CR761983.1 | - | 1 - 2 | 587 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |