Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008025.1 | NP_001008026.1 | 4 - 5 | 505 | 107 | |
GenBank mRNA | BC080905.1 | - | 4 - 5 | 505 | 107 | |
CR926163.1 | - | 4 - 5 | 480 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |