Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008098.2 | NP_001008099.1 | 2 - 3 | 193 | 99 | |
XM_012966474.1 | XP_012821928.1 | 1 - 2 | 456 | 99 | ||
XM_012966475.1 | XP_012821929.1 | 3 - 4 | 664 | 99 | ||
GenBank mRNA | BC081300.1 | - | 2 - 3 | 197 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |