Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008204.1 | NP_001008205.1 | 9 - 10 | 965 | 81 | |
XM_012968277.2 | XP_012823731.1 | 9 - 10 | 941 | 81 | ||
XM_012968278.2 | XP_012823732.1 | 9 - 10 | 1022 | 81 | ||
GenBank mRNA | BC080963.1 | - | 9 - 10 | 965 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |