Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008431.2 | NP_001008431.1 | 5 - 6 | 1287 | 82 | |
GenBank mRNA | BC080136.1 | - | 5 - 6 | 1290 | 82 | |
CR761999.2 | - | 5 - 6 | 931 | 82 | ||
CR848157.2 | - | 5 - 6 | 1255 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |