Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011071.1 | NP_001011071.1 | 6 - 7 | 657 | 101 | |
XM_012953486.2 | XP_012808940.1 | 6 - 7 | 638 | 101 | ||
GenBank mRNA | BC084454.1 | - | 6 - 7 | 657 | 101 | |
CR761781.2 | - | 5 - 6 | 565 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |