Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011113.2 | NP_001011113.1 | 3 - 4 | 282 | 80 | |
GenBank mRNA | BC084511.1 | - | 3 - 4 | 284 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |