Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp4r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011151.1 | NP_001011151.1 | 5 - 6 | 612 | 65 | |
GenBank mRNA | BC084901.1 | - | 5 - 6 | 612 | 65 | |
CR760404.1 | - | 5 - 6 | 498 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |