Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011225.1 | NP_001011225.1 | 5 - 6 | 389 | 100 | |
XM_012960232.1 | XP_012815686.1 | 5 - 6 | 481 | 100 | ||
XM_012960233.1 | XP_012815687.1 | 5 - 6 | 466 | 100 | ||
XM_012960234.2 | XP_012815688.1 | 5 - 6 | 477 | 100 | ||
XM_012960235.1 | XP_012815689.1 | 5 - 6 | 481 | 100 | ||
XM_012960236.2 | XP_012815690.1 | 2 - 3 | 153 | 100 | ||
GenBank mRNA | BC087789.1 | - | 5 - 6 | 389 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |