Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtpbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011239.1 | NP_001011239.1 | 2 - 3 | 288 | 69 | |
GenBank mRNA | BC087811.1 | - | 2 - 3 | 288 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |