Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011270.1 | NP_001011270.1 | 3 - 4 | 286 | 110 | |
XM_012958933.2 | XP_012814387.1 | 4 - 5 | 692 | 110 | ||
XM_012958934.2 | XP_012814388.1 | 3 - 4 | 499 | 110 | ||
XM_012958935.2 | XP_012814389.1 | 3 - 4 | 1226 | 110 | ||
XM_012958937.2 | XP_012814391.1 | 2 - 3 | 143 | 110 | ||
XM_018092056.1 | XP_017947545.1 | 4 - 5 | 688 | 110 | ||
GenBank mRNA | BC087979.1 | - | 3 - 4 | 286 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |