Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011274.1 | NP_001011274.1 | 3 - 4 | 659 | 54 | |
GenBank mRNA | BC087986.1 | - | 3 - 4 | 659 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |