Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011185.2 | NP_001011185.1 | 5 - 6 | 441 | 103 | |
GenBank mRNA | BC086504.2 | - | 5 - 6 | 441 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |