Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atraid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011316.1 | NP_001011316.1 | 571 | 71 | ||
GenBank mRNA | BC088051.1 | - | 571 | 71 | ||
CU075626.1 | - | 4 - 5 | 567 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |