Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011378.1 | NP_001011378.1 | 3 - 4 | 263 | 71 | |
GenBank mRNA | BC088559.1 | - | 3 - 4 | 263 | 71 | |
CR855398.2 | - | 3 - 4 | 257 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |