Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zmynd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011379.1 | NP_001011379.1 | 4 - 5 | 648 | 98 | |
GenBank mRNA | BC088560.1 | - | 4 - 5 | 648 | 98 | |
CR848399.2 | - | 4 - 5 | 651 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |