Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gspt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015805.2 | NP_001015805.2 | 11 - 12 | 1685 | 71 | |
GenBank mRNA | BC064719.1 | - | 11 - 12 | 1617 | 71 | |
BC090081.1 | - | 11 - 12 | 1589 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |