Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015811.1 | NP_001015811.1 | 1 - 2 | 368 | 58 | |
GenBank mRNA | BC090092.1 | - | 1 - 2 | 368 | 58 | |
CR762212.2 | - | 1 - 2 | 282 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |