Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015845.1 | NP_001015845.1 | 14 - 15 | 1448 | 69 | |
GenBank mRNA | BC090134.1 | - | 14 - 15 | 1448 | 69 | |
BC122899.1 | - | 14 - 15 | 1405 | 69 | ||
CR848552.2 | - | 14 - 15 | 1455 | 69 |
Gene Symbol: LOC101731871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004911912.3 | XP_004911969.1 | 14 - 15 | 1499 | 69 | |
XM_004911913.3 | XP_004911970.1 | 14 - 15 | 1492 | 69 | ||
XM_004911914.3 | XP_004911971.1 | 14 - 15 | 1465 | 69 | ||
XM_012957841.2 | XP_012813295.1 | 14 - 15 | 1458 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |