Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cmtm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015861.1 | NP_001015861.1 | 2 - 3 | 360 | 100 | |
GenBank mRNA | BC090372.1 | - | 2 - 3 | 360 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |