Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015903.1 | NP_001015903.1 | 6 - 7 | 768 | 89 | |
GenBank mRNA | BC121632.1 | - | 5 - 6 | 598 | 89 | |
BC161440.1 | - | 6 - 7 | 776 | 89 | ||
BC171009.1 | - | 6 - 7 | 724 | 89 | ||
CR926439.1 | - | 6 - 7 | 768 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |