Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015940.2 | NP_001015940.1 | 3 - 4 | 888 | 110 | |
GenBank mRNA | CR848495.2 | - | 3 - 4 | 888 | 110 | |
CT030375.1 | - | 3 - 4 | 879 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |