Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3h15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016105.2 | NP_001016105.1 | 9 - 10 | 1208 | 99 | |
GenBank mRNA | BC135295.1 | - | 9 - 10 | 1129 | 99 | |
CR762127.2 | - | 9 - 10 | 1208 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |