Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016197.2 | NP_001016197.1 | 5 - 6 | 988 | 82 | |
XM_012966564.2 | XP_012822018.1 | 5 - 6 | 1531 | 82 | ||
GenBank mRNA | BC170733.1 | - | 4 - 5 | 957 | 82 | |
BC170735.1 | - | 4 - 5 | 957 | 82 | ||
CR926348.2 | - | 5 - 6 | 988 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |