Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016202.2 | NP_001016202.1 | 13 - 14 | 1632 | 56 | |
XM_012970406.2 | XP_012825860.1 | 14 - 15 | 1831 | 56 | ||
XM_012970407.2 | XP_012825861.1 | 14 - 15 | 1816 | 56 | ||
XM_012970408.2 | XP_012825862.1 | 13 - 14 | 1754 | 56 | ||
XM_012970409.2 | XP_012825863.1 | 12 - 13 | 1635 | 56 | ||
XM_012970410.2 | XP_012825864.1 | 12 - 13 | 1563 | 56 | ||
GenBank mRNA | BC091592.1 | - | 13 - 14 | 1643 | 56 | |
CR926343.2 | - | 13 - 14 | 1632 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |