Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016366.2 | NP_001016366.1 | 5 - 6 | 638 | 80 | |
XM_012961090.2 | XP_012816544.1 | 5 - 6 | 682 | 80 | ||
GenBank mRNA | BC153695.1 | - | 4 - 5 | 548 | 80 | |
CR761286.2 | - | 5 - 6 | 638 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |