Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016428.2 | NP_001016428.1 | 5 - 6 | 628 | 97 | |
GenBank mRNA | BC171075.1 | - | 5 - 6 | 582 | 97 | |
BC171077.1 | - | 5 - 6 | 582 | 97 | ||
CR761059.2 | - | 5 - 6 | 628 | 97 | ||
CR761251.2 | - | 336 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |