Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016534.1 | NP_001016534.1 | 4 - 5 | 359 | 99 | |
XM_018094654.1 | XP_017950143.1 | 5 - 6 | 759 | 99 | ||
GenBank mRNA | CR926169.1 | - | 4 - 5 | 359 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |