Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016558.1 | NP_001016558.1 | 2 - 3 | 391 | 98 | |
GenBank mRNA | CR760702.1 | - | 2 - 3 | 391 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |