Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016835.2 | NP_001016835.1 | 5 - 6 | 905 | 89 | |
XM_012957004.2 | XP_012812458.1 | 5 - 6 | 1338 | 89 | ||
GenBank mRNA | BC123045.1 | - | 5 - 6 | 1127 | 89 | |
CR855489.2 | - | 5 - 6 | 905 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |