Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016845.2 | NP_001016845.1 | 2 - 3 | 250 | 65 | |
XM_012962938.1 | XP_012818392.1 | 3 - 4 | 296 | 65 | ||
GenBank mRNA | BC158298.1 | - | 2 - 3 | 261 | 65 | |
CR855446.2 | - | 2 - 3 | 250 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |