Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016870.2 | NP_001016870.1 | 3 - 4 | 381 | 115 | |
GenBank mRNA | BC135749.1 | - | 3 - 4 | 426 | 115 | |
CR848627.2 | - | 3 - 4 | 381 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |