Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017294.2 | NP_001017294.1 | 202 | 76 | ||
GenBank mRNA | BC158374.1 | - | 1 - 2 | 206 | 76 | |
CR855640.2 | - | 202 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |