Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017303.2 | NP_001017303.1 | 1 - 2 | 126 | 69 | |
GenBank mRNA | BC152034.1 | - | 1 - 2 | 139 | 69 | |
CR855571.2 | - | 1 - 2 | 126 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |