Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030393.1 | NP_001025564.1 | 4 - 5 | 390 | 84 | |
GenBank mRNA | BC090604.1 | - | 4 - 5 | 390 | 84 | |
CT030593.1 | - | 4 - 5 | 368 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |