Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030449.1 | NP_001025620.1 | 326 | 94 | ||
GenBank mRNA | BC091085.1 | - | 326 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |