Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030479.1 | NP_001025650.1 | 6 - 7 | 902 | 102 | |
GenBank mRNA | BC091643.1 | - | 6 - 7 | 902 | 102 | |
CR942452.2 | - | 6 - 7 | 939 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |