Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032339.1 | NP_001027510.1 | 2 - 3 | 173 | 76 | |
XM_012963846.2 | XP_012819300.1 | 2 - 3 | 318 | 76 | ||
XM_018093866.1 | XP_017949355.1 | 2 - 3 | 319 | 76 | ||
GenBank mRNA | BC098189.1 | - | 2 - 3 | 173 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |