Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007910.2 | NP_001007911.1 | 4 - 5 | 490 | 64 | |
GenBank mRNA | BC080349.1 | - | 4 - 5 | 450 | 64 | |
CR760684.2 | - | 4 - 5 | 490 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |