Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008064.2 | NP_001008065.2 | 4 - 5 | 690 | 107 | |
GenBank mRNA | BC080953.1 | - | 4 - 5 | 737 | 107 | |
CR848500.2 | - | 4 - 5 | 690 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |