Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011430.2 | NP_001011430.2 | 4 - 5 | 589 | 80 | |
GenBank mRNA | BC088067.1 | - | 3 - 4 | 276 | 80 | |
CR761204.2 | - | 4 - 5 | 589 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |