Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc39a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015957.2 | NP_001015957.1 | 1 - 2 | 159 | 99 | |
GenBank mRNA | BC121996.1 | - | 1 - 2 | 136 | 99 | |
CR848459.2 | - | 1 - 2 | 159 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |