Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015961.2 | NP_001015961.1 | 3 - 4 | 705 | 94 | |
GenBank mRNA | BC122010.1 | - | 3 - 4 | 577 | 94 | |
BC161506.1 | - | 3 - 4 | 560 | 94 | ||
CR848436.2 | - | 3 - 4 | 705 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |