Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015994.2 | NP_001015994.1 | 2 - 3 | 219 | 92 | |
XM_018093034.1 | XP_017948523.1 | 3 - 4 | 304 | 92 | ||
GenBank mRNA | BC167873.1 | - | 2 - 3 | 252 | 92 | |
CR848305.2 | - | 2 - 3 | 219 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |