Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016049.2 | NP_001016049.1 | 536 | 101 | ||
GenBank mRNA | BC161482.1 | - | 4 - 5 | 533 | 101 | |
BC170571.1 | - | 533 | 101 | |||
BC170573.1 | - | 533 | 101 | |||
CR762349.2 | - | 536 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |