Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016112.2 | NP_001016112.1 | 8 - 9 | 790 | 69 | |
XM_012961997.2 | XP_012817451.1 | 9 - 10 | 898 | 69 | ||
XM_012961998.2 | XP_012817452.1 | 9 - 10 | 894 | 69 | ||
GenBank mRNA | BC121530.1 | - | 9 - 10 | 842 | 69 | |
CR762106.2 | - | 8 - 9 | 790 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |